missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIGLEC H Monoclonal Antibody (eBio440c), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
150.00€ - 395.00€
Spécification
Antigène | SIGLEC H |
---|---|
Clone | eBio440c |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16371144
|
Affymetrix eBioscience
62-0333-82 |
100μg |
395.00€
100µg |
Expédition estimée: 10-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16361144
|
Affymetrix eBioscience
62-0333-80 |
25 μg |
Cet article est actuellement indisponible ou discontinu.
Veuillez consulter la page produit pour de possible alternative dans l’onglet |
||||||
Description
The monoclonal antibody eBio440c recognizes Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. Mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+
Siglec-H, a protein exclusively found on pDC (plasmacytoid dendritic cells) or type I IFN-producing cells (IPC) in the naive mouse. Mouse IPC are typically PDCA+, CD11c+, CD11b-, B220+, and Ly-6C+, and are quick to respond to viruses. Siglec-H is a transmembrane protein of the Ig superfamily that like CD33 have been shown to bind sialic acid but lacks the characteristic cytoplasmic ITIM domain (immunoreceptor tyrosine based inhibitory motif). To overcome the lack of a cytoplasmic domain, Siglec-H associates with DAP12 thereby allowing for signal transduction. The eBio440c antibody has been shown to inhibit pDC function (inhbits IFNalpha secretion in response to CpG). It has been observed that some mouse strains (such as SJL) have higher percentages of pDCs compared to C57BL/6.Spécification
SIGLEC H | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
233274 | |
IgG2b κ | |
4° C, store in dark, DO NOT FREEZE! | |
Siglech |
eBio440c | |
Flow Cytometry | |
Super Bright 436 | |
Rat | |
Mouse | |
Siglech | |
Primary | |
Antibody |