missing translation for 'onlineSavingsMsg'
                
Learn More
Learn More
CSF1R (MCSFR) Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™
Code nomenclature Nacres: NA.28
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                :
                            
                        
                        
                            
                                50ug
                            
                        
                    Description
Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies. 
                        
                        
                        
                        
                        
                        
                        
                        
                            - Activity: Measured by its ability to bind recombinant mouse IL34 in a functional ELISA; immobilized mouse CSF1R-His at 10μg/mL (100μL/well) can bind biotinylated human CSF1-His; the ED50 of biotinylated human CSF1-His is 0.50μg/mL
 - Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
 - Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
 
Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.
            Spécification
Spécification
| À utiliser avec (application) | Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | 
| Identification génétique (Entrez) | 12978 | 
| Poids moléculaire | 56.8kDa | 
| Quantité | 50 μg | 
| Conditions de stockage | Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | 
| Forme structurelle | Recombinant/Cell Surface Protein | 
| Concentration en endotoxines | <1 EU/μg | 
| Alias de gène | ICsf1r, AI323359, CD115, CSF-1R, Csfmr, Fim-2, Fms, M-CSFR | 
| Nom usuel | Colony stimulating factor 1 receptor | 
| Symbole de gène(s) | CSF1R | 
| Afficher plus | 
For Research Use Only. Not for use in diagnostic procedures.
                    Correction du contenu d'un produit
                
                Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
			CSF1R (MCSFR) Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™ > 
            Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu