missing translation for 'onlineSavingsMsg'
Learn More

Sirtuin 1/SIRT1 Rabbit anti-Human, Clone: 12, Novus Biologicals™

Rabbit Monoclonal Antibody

Marque:  Novus Biologicals NBP2-89977-50UL

Code nomenclature Nacres: NA.46

 Afficher plus de versions de ce produit

Code produit. 18225958

  • 367.00€ / 50µL

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Description

Description

Sirtuin 1/SIRT1 Monoclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Spécification

Spécification

Sirtuin 1/SIRT1
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
ELISA
12
ELISA 1:5000-1:10000
EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1
Recombinant Human Sirtuin 1/SIRT1 protein
50 μg
Apoptosis, Chromatin Research, Cognition and Behavior, DNA Repair, Epigenetics, Phospho Specific
23411
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.