Learn More
SLC5A3 Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Marque: Invitrogen PA19554
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
PA1-9554 detects SLC5A3 in human samples. PA1-9554 has been successfully used in Western blot procedures. PA1-9554 immunogen is a purified recombinant SLC5A3 corresponding to the chromosome 21 gene (Human).
Prevents intracellular accumulation of high concentrations of myo-inositol (an osmolyte) that result in impairment of cellular function.Spécification
SLC5A3 | |
Polyclonal | |
Unconjugated | |
SLC5A3 | |
AA623876; BCW2; BCW2 protein; BF642829; Na(+)/myo-inositol cotransporter; Slc5a3; SMIT; Smit1; SMIT2; sodium myo-inositol transporter 1; sodium/myo-inositol cotransporter; sodium/myo-inositol cotransporter 1; sodium/myo-inositol transporter 1; solute carrier family 5 (inositol transporters), member 3; solute carrier family 5 (sodium/myo-inositol cotransporter), member 3; solute carrier family 5 member 3; solute carrier family 5, member 3; solute carrier family 5, member alpha 3 | |
Chicken | |
IgY fraction | |
RUO | |
6526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P53794 | |
SLC5A3 | |
Purified recombinant SLC5A3 corresponding to the chromosome 21 gene (Human). | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgY |