missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMN Mouse anti-Human, Clone: 2C6D9, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60154-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This antibody 60154-1-Ig is specific to human SMN2. It can't recognize mouse and rat SMN.
The StARD (steroidogenic acute regulatory protein-related lipid transfer (START) domain containing) family of proteins is comprised of fifteen different members. All members contain the characteristic START domain and are believed to play key roles in the metabolism and transport of lipids. The StARD proteins are grouped into six subfamilies based on their START domain sequences. PC-TP (StARD2), StARD7, StARD10 and GPBP (StARD11) constitute one subfamily, namely the STARD2/PCTP group. StARD10, also known as PCTP2, PCTPL, NY-CO-28, CGI-52 or SDCCAG28, is widely expressed and functions in phospholipid transfer, binding to phosphatidylcholine and phosphatidylethanolamine. StARD10 can be found in sperm flagellum, potentially functioning as an enzyme involved in energy metabolism, and its expression is developmentally regulated in testis and mammary glands. StARD10 activity can be inhibited via phosphorylation by casein kinase II.Spécification
SMN | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16637 | |
SMN1, SMN2 | |
SMN (Human-Specific) Fusion Protein Ag14333 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
2C6D9 | |
Unconjugated | |
SMN2 | |
C BCD541, Component of gems 1, Gemin 1, SMN, SMN1, SMN1,SMN, SMN2, SMNC, SMNT, Survival motor neuron protein | |
Mouse | |
Protein A | |
RUO | |
6606, 6607 | |
-20°C | |
Liquid |