missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNAI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13099-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.Spécification
SNAI1 | |
Polyclonal | |
Unconjugated | |
SNAI1 | |
Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAI1, snail homolog 1 (Drosophila), SNAIL1, Zinc finger protein SNAI1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
116490, 20613, 6615 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95863, Q02085 | |
SNAI1 | |
SNAI1 Fusion Protein Ag3723 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |