missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SOD2/Mn-SOD Antibody (8H3D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-61883-0.025ml
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SOD2/Mn-SOD Monoclonal specifically detects SOD2/Mn-SOD in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| SOD2/Mn-SOD | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| EC 1.15.1.1, indophenoloxidase B, IPOB, manganese-containing superoxide dismutase, mangano-superoxide dismutase, Mn superoxide dismutase, MNSOD, MVCD6, superoxide dismutase [Mn], mitochondrial, superoxide dismutase 2, mitochondrial | |
| Mouse | |
| 88 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| 8H3D2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| SOD2 | |
| Purified recombinant fragment of human SOD2 (AA: 1-222) expressed in E. Coli. | |
| Protein G purified | |
| Cancer, Diabetes Research, Neuroscience, Tumor Suppressors | |
| 6648 | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu