missing translation for 'onlineSavingsMsg'
Learn More

SPANXC Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 11957-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829373

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family, which is located in a gene cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a protein that localizes to the nucleus and is expressed in highly metastatic cell lines, making the protein a potential diagnostic and prognostic marker. The protein belongs to a family of cancer/testis antigens and represents a potential target for cancer immunotherapy.
Spécification

Spécification

SPANXC
Polyclonal
Unconjugated
SPANXC
C, Cancer/testis antigen 11.3, CT11.3, CTp11, SPANX C, SPANX family member C, SPANX family, member C, SPANXC
Rabbit
Antigen Affinity Chromatography
RUO
64663
-20°C
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NY87
SPANXC
SPANXC Fusion Protein Ag2561
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions