missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spartin/SPG20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13791-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
SPG20 is a protein containing a MIT (Microtubule Interacting and Trafficking molecule) domain, and is implicated in regulating endosomal trafficking and mitochondria function. The protein localizes to mitochondria and partially co-localizes with microtubules. Stimulation with epidermal growth factor (EGF) results in protein translocation to the plasma membrane, and the protein functions in the degradation and intracellular trafficking of EGF receptor.Spécification
Spartin/SPG20 | |
Polyclonal | |
Unconjugated | |
SPART | |
KIAA0610, SPARTIN, Spastic paraplegia 20 protein, SPG20, TAHCCP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
229285, 23111, 295053 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N0X7, Q8R1X6 | |
Spart, Spg20 | |
Spartin, SPG20 Fusion Protein Ag4815 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |