missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPERT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17826-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
SPERT (spermatid associated protein), also known as CBY2 (chibby homolog 2) or NURIT, is a novel leucine-zipper protein belonging to the chibby family of proteins. Uniquely Expressed in the spermatid flower-like structure, SPERT interacts with Nek1, a member of the NIMA-family kinase family that is associated centrosomal stability and ciliogenesis. It contains a leucine-zipper motif and two coiled-coil regions, and is transcribed through the elongation stage of the spermatids. SPERT is absent from mature spermatozoa and is thought to be involved in transporting proteins that are to be discarded via the residual bodies.Spécification
SPERT | |
Polyclonal | |
Unconjugated | |
SPERT | |
CBY2, NURIT, Protein chibby homolog 2, spermatid associated, Spermatid associated protein, SPERT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
220082, 498572, 67926 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q32MG2, Q6AXV6, Q8NA61 | |
Spert | |
SPERT Fusion Protein Ag12200 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |