missing translation for 'onlineSavingsMsg'
Learn More

ST3GAL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13620-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831924

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

ST3GAL2 is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A. The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP-sialic acid to galactose-containing substrates. The encoded protein is normally found in the Golgi but can be proteolytically processed to a soluble form. This protein, which is a member of glycosyltransferase family 29, can use the same acceptor substrates as does sialyltransferase 4A.
Spécification

Spécification

ST3GAL2
Polyclonal
Unconjugated
ST3GAL2
Alpha 2,3 ST 2, Gal NAc6S, Sialyltransferase 4B, SIAT4 B, SIAT4B, ST3Gal II, ST3GAL2, ST3GalA.2, ST3GALII
Rabbit
Antigen Affinity Chromatography
RUO
20444, 64442, 6483
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.14 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q11205, Q16842
St3gal2
ST3GAL2 Fusion Protein Ag4528
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions