missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STBD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11842-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
May have the capability to bind to carbohydrates.Spécification
STBD1 | |
Polyclonal | |
Unconjugated | |
STBD1 | |
Genethonin 1, GENEX3414, GENX 3414, starch binding domain 1, STBD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52331, 8987 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95210, Q8C7E7 | |
Stbd1 | |
STBD1 Fusion Protein Ag2481 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |