missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRA6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22001-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
STRA6 is a membrane protein involved in the metabolism of retinol. The encoded protein acts as a receptor for retinol/retinol binding protein complexes. This protein removes the retinol from the complex and transports it across the cell membrane. Defects in this gene are a cause of syndromic microphthalmia type 9 (MCOPS9).Spécification
STRA6 | |
Polyclonal | |
Unconjugated | |
STRA6 | |
MCOPS9, PP14296, STRA6, UNQ3126/PRO10282/PRO19578 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20897, 64220 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70491, Q9BX79 | |
STRA6 | |
STRA6 Fusion Protein Ag17591 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |