missing translation for 'onlineSavingsMsg'
Learn More

STXBP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14231-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832774

  • 121.38€ / 20µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

STXBP4 plays a role in the translocation of transport vesicles from the cytoplasm to the plasma membrane. STXBP4 inhibits the translocation of SLC2A4 from intracellular vesicles to the plasma membrane by STX4A binding and preventing the interaction between STX4A and VAMP2. Stimulation with insulin disrupts the interaction with STX4A, leading to increased levels of SLC2A4 at the plasma membrane. STXBP4 may also play a role in the regulation of insulin release by pancreatic beta cells after stimulation by glucose.
Spécification

Spécification

STXBP4
Polyclonal
Unconjugated
STXBP4
STX4 interacting protein, STXBP4, Synip, Syntaxin 4 interacting protein, syntaxin binding protein 4
Rabbit
Antigen Affinity Chromatography
RUO
20913, 252983, 303443
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6ZWJ1, Q9WV89
STXBP4
STXBP4 Fusion Protein Ag5471
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions