missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1B1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16050-1-AP-20UL
Additional Details : Poids : 0.00010kg
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure is similar among family members. However, the total genomic length of this gene is greater than that of other SULT1 genes.Specifications
SULT1B1 | |
Polyclonal | |
Unconjugated | |
SULT1B1 | |
ST1B1, ST1B2, Sulfotransferase 1B1, Sulfotransferase 1B2, SULT1B1, SULT1B2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27284 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43704 | |
SULT1B1 | |
SULT1B1 Fusion Protein Ag9009 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |