missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-56031-25ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Syntaxin 1A Polyclonal specifically detects Syntaxin 1A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| Syntaxin 1A | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| STX1A | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:QRQLEITGRTTTSEELEDMLESGNPAIF | |
| 25 μL | |
| Membrane Trafficking and Chaperones, Neuronal Cell Markers, Neuroscience, Signal Transduction | |
| 6804 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu