missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15556-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.Spécification
Syntaxin 3 | |
Polyclonal | |
Unconjugated | |
STX3 | |
STX3, STX3A, syntaxin 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20908, 6809, 81802 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08849, Q13277, Q64704 | |
Stx3 | |
Syntaxin 3 Fusion Protein Ag7925 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |