missing translation for 'onlineSavingsMsg'
Learn More

Syntaxin 7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 12322-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829973

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
Spécification

Spécification

Syntaxin 7
Polyclonal
Unconjugated
STX7
STX7, syntaxin 7
Rabbit
Antigen Affinity Chromatography
RUO
53331, 60466, 8417
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.4 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O15400, O70257, O70439
Stx7
Syntaxin 7 Fusion Protein Ag2975
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions