Learn More
αβ T-Cell Receptor Mouse anti-Rat, FITC, Clone: R73, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 554913
Description
The R73 antibody reacts with the αβ T-cell Receptor (TCR) found on most peripheral T lymphocytes, intestinal intraepithelial lymphocytes, and thymocytes. It does not react with γδ TCR-bearing cells. Cross-linked R73 mAb induces T-cell differentiation and activation. In vivo treatment with mAb R73 can suppress immune function of peripheral αβ TCR-expressing T cells, and reduce the severity of experimental autoimmune, transplant rejection, and graft-versus-host responses.
Spécification
| αβ T-Cell Receptor | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat T blasts and rat erythrocytes | |
| 0.5 mg | |
| Immunology | |
| Rat | |
| IgG1 κ |
| Flow Cytometry | |
| R73 | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only.