Learn More
Tensin 4/CTEN Antibody (S06-1F3), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-19791-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Tensin 4/CTEN Monoclonal antibody specifically detects Tensin 4/CTEN in Human, Mouse, Rat samples. It is validated for Western Blot
Spécification
| Tensin 4/CTEN | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84951 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| S06-1F3 | |
| Unconjugated | |
| CTENC terminal tensin like, C-terminal tensin-like protein, FLJ14950, tensin 4, tensin-4 | |
| A synthetic peptide of human Tensin 4/CTEN (Uniprot # Q8IZW8) | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |