missing translation for 'onlineSavingsMsg'
Learn More

TFAM Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23996-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832595

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

TMEM192 is a widely expressed, four transmembrane protein that localizes to late endosomal/lysosomal membranes. This localization is mediated by two dileucine motifs present at the amino-terminus of TMEM192. A cysteine residue near the carboxy residue is responsible for disulfide bond formation in TMEM192 homodimers. Little is known about the function of this protein.
Spécification

Spécification

TFAM
Polyclonal
Unconjugated
TFAM
MtTF1, mtTFA, TCF 6, TCF6, TCF6L1, TCF6L2, TCF6L3, TFAM, Transcription factor 6, Transcription factor 6 like 2
Rabbit
Antigen Affinity Chromatography
RUO
7019
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q00059
TFAM
TFAM Fusion Protein Ag21196
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions