missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin / CD141 (Endothelial Cell Marker) Mouse, Clone: rTHBD/1591, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 7056-MSM3-P0
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Specifications
| Thrombomodulin / CD141 (Endothelial Cell Marker) | |
| Cell Surface and Cytoplasmic | |
| rTHBD/1591 | |
| Unconjugated | |
| Mouse | |
| ∽100kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 7056 | |
| IgG1 |
| ELISA, Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant human Thrombomodulin protein fragment (apprx. aa 69-194) | |
| THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction