missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thyroglobulin Mouse anti-Human, Mouse, Rat, Clone: rTGB/4744, Novus Biologicals™
Mouse Monoclonal Antibody
499.00€
Spécification
| Antigène | Thyroglobulin |
|---|---|
| Clone | rTGB/4744 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
Thyroglobulin Monoclonal specifically detects Thyroglobulin in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.Spécification
| Thyroglobulin | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cell Biology | |
| 10 mM PBS with 0.05% BSA | |
| 7038 | |
| IgG2a κ | |
| Protein A or G purified |
| rTGB/4744 | |
| Flow Cytometry 0.5-1 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AITD3TGN, TDH3, Tg, thyroglobulin | |
| Human thyroid follicular cells | |
| Primary | |
| Store at 4°C. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit