Learn More
TIAR Mouse anti-Human, Unconjugated, Clone: 1E8D12, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66907-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The TIAR antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TIAR. This antibody recognizes human, mouse, rat antigen. The TIAR antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
| TIAR | |
| Monoclonal | |
| 940 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1E8D12 | |
| Unconjugated | |
| 20 μL | |
| 7073 | |
| IgG1 |