missing translation for 'onlineSavingsMsg'
Learn More

TIMELESS Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14421-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833054

  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Timeless is required for normal progression of S-phase. Involved in the circadian rhythm autoregulatory loop. Negatively regulates CLOCK-NPAS2/BMAL1-induced transactivation of PER1 possibly via translocation of PER1 into the nucleus. Promotes TIPIN nuclear localization. Involved in cell survival after DNA damage or replication stress. May be specifically required for the ATR-CHK1 pathway in the replication checkpoint induced by hydroxyurea or ultraviolet light. May also play an important role in epithelial cell morphogenesis and formation of branching tubules.
Spécification

Spécification

TIMELESS
Polyclonal
Unconjugated
TIMELESS
hTIM, Protein timeless homolog, TIM, TIM1, TIMELESS, timeless homolog (Drosophila), TIMELESS1
Rabbit
Antigen Affinity Chromatography
RUO
8914
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UNS1
TIMELESS
TIMELESS Fusion Protein Ag5767
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions