missing translation for 'onlineSavingsMsg'
Learn More

TIPIN Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications

433.00€ - 624.00€

Spécification

Antigène TIPIN
Dilution Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugué Unconjugated
View More Specs

Products 2
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
18427930
View Documents
Novus Biologicals
NBP1-83650-25ul
25 μL
433.00€
25µL
View alternative products
Save Up to 
»
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
18291226
View Documents
Novus Biologicals
NBP1-83650
0.1 mL
624.00€
0.10mL
View alternative products
Save Up to 
»
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

TIPIN Polyclonal specifically detects TIPIN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TIPIN
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
RUO
Human
FLJ20516, TIMELESS interacting protein, TIMELESS-interacting protein
TIPIN
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000
Polyclonal
Rabbit
DNA Repair
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
54962
This antibody was developed against Recombinant Protein corresponding to amino acids:EEQQQRIERNKQLALERRQAKLLSNSQTLGNDMLMNTPRAHTVEEVNTDEDQKEESNGLNEDILDNPCNDAIA
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.