missing translation for 'onlineSavingsMsg'
Learn More

TLR5 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 19810-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16829294

  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

TLR5 is a Type I membrane protein belonging to the Toll-like receptor family, participates in the innate immune response to microbial agents. TLR5 also plays a role in mediating detection of bacterial flagellins. TLR5 acts via MyD88 and TRAF6, leading to NF-kappa-B activation, cytokine secretion and the inflammatory response. TLR5 protein binds to TIRAP and MyD88 via their respective TIR domains TLR5 is highly expressed in liver, and is detected in lung and at very low levels in most other tissues. The TLR5 gene lies in a locus that is associated with susceptibility to Salmonella. Inbred strains of mice can be classified into 3 categories according to their resistance to infection with S.typhimurium: susceptible (BALB/c, C57BL/6, C3H/He), intermediate (DBA/2, C75L) and resistant (A, CBA). The strain MOLF/Ei is highly susceptible to the infection, has a unique TLR5 haplotype and a lower expression of TRL5. A common TLR5 stop codon polymorphism that disrupts TLR5 signaling is associated with susceptibility to Legionnaires′ disease and demonstrates the importance of TLR5 in the innate immune response.
Specifications

Specifications

TLR5
Polyclonal
Unconjugated
TLR5
SLEB1, TIL3, TLR5, toll like receptor 5
Rabbit
Antigen Affinity Chromatography
RUO
53791, 7100
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60602, Q9JLF7
Tlr5
TLR5 Fusion Protein Ag13840
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers