Learn More
TNFAIP2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 25649-1-AP-150UL
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17241376
  • 399.00€ / 150µL
Expédition estimée: 27-05-2024
Description
TNFAIP2, also named as B94, belongs to the SEC6 family. TNFAIP2 may play a role as a mediator of inflammation and angiogenesis. TNFAIP2 is differentially expressed in development and capillary tube-like formation in vitro. It is induced by TNFα and other proinflammatory factors.
Spécification
Spécification
TNFAIP2
Polyclonal
Unconjugated
150 μL
7127
Liquid
Western Blot, ELISA
700 μg/mL