Learn More
TNFAIP2 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25649-1-AP-150UL
Informations supplémentaires : Poids : 0.00020kg
Description
TNFAIP2, also named as B94, belongs to the SEC6 family. TNFAIP2 may play a role as a mediator of inflammation and angiogenesis. TNFAIP2 is differentially expressed in development and capillary tube-like formation in vitro. It is induced by TNFα and other proinflammatory factors.
Spécification
TNFAIP2 | |
Polyclonal | |
Unconjugated | |
150 μL | |
7127 | |
Liquid |
Western Blot, ELISA | |
700 μg/mL | |
Rabbit | |
Primary | |
Human |