missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNRC4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25353-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
RNA-binding protein involved in the regulation of pre- mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of cardiac isoforms of TNNT2 during heart remodeling at the juvenile to adult transition. Activates the splicing of MAPT/Tau exon 10. Binds to muscle-specific splicing enhancer (MSE) intronic sites flanking the alternative exon 5 of TNNT2 pre-mRNA.Spécification
TNRC4 | |
Polyclonal | |
Unconjugated | |
CELF3 | |
Bruno like protein 1, BRUNOL1, CAG repeat protein 4, CAGH4, CELF 3, CELF3, ERDA4, ETR 1, RNA binding protein BRUNOL 1, TNRC4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11189, 78784 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5SZQ8, Q8CIN6 | |
Celf3 | |
TNRC4 Fusion Protein Ag18601 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |