missing translation for 'onlineSavingsMsg'
Learn More

TPMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10682-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16837223

  • 140.00€ / 20µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody specifically recognizes the 34 kDa protein.

Catalyzes the S-methylation of thiopurine drugs such as 6-mercaptopurine. Defects in TPMT are the cause of thiopurine S-methyltransferase deficiency (TPMT deficiency). TPMT is an enzyme involved in the normal metabolic inactivation of thiopurine drugs. These drugs are generally used as immunosupressants or cytotoxic drugs and are prescribed for a variety of clinical conditions including leukemia, autoimmune disease and organ transplantation. Patients with intermediate or no TPMT activity are at risk of toxicity after receiving standard doses of thiopurine drugs and it is shown that inter-individual differences in response to these drugs are largely determined by genetic variation at the TPMT locus.
Spécification

Spécification

TPMT
Polyclonal
Unconjugated
TPMT
Thiopurine methyltransferase, thiopurine S methyltransferase, TPMT
Rabbit
Antigen Affinity Chromatography
RUO
22017, 690050, 7172
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin)
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O55060, P51580, Q9Z0T0
TPMT
TPMT Fusion Protein Ag1074
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions