Learn More
TRAIL Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27064-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The TRAIL antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human TRAIL. This antibody recognizes human antigen. The TRAIL antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
TRAIL | |
Polyclonal | |
Unconjugated | |
150 μL | |
8743 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
333 μg/mL | |
Rabbit | |
Primary | |
Human |