missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13311-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Necessary for collagen type I synthesis. May couple the activity of the ER Ca(2+) pump SERCA2B with the activity of the translocon. This coupling may increase the local Ca(2+) concentration at the site of collagen synthesis, and a high Ca(2+) concentration may be necessary for the function of molecular chaperones involved in collagen folding. Required for proper insertion of the first transmembrane helix N-terminus of TM4SF20 into the ER lumen, may act as a ceramide sensor for regulated alternative translocation (RAT).Spécification
TRAM2 | |
Polyclonal | |
Unconjugated | |
TRAM2 | |
KIAA0057, TRAM2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100361830, 170829, 9697 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15035, Q924Z5 | |
Tram2 | |
TRAM2 Fusion Protein Ag4144 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |