missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27599-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).Spécification
TREM2 | |
Polyclonal | |
Unconjugated | |
TREM2 | |
TREM 2, TREM2, Trem2a, Trem2b, Trem2c | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54209 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NZC2 | |
TREM2 | |
TREM2 Fusion Protein Ag26319 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |