missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIB2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15359-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes one of three members of the Tribbles family. The Tribbles members share a Trb domain, which is homologous to protein serine-threonine kinases, but lacks the active site lysine and probably lacks a catalytic function. The Tribbles proteins interact and modulate the activity of signal transduction pathways in a number of physiological and pathological processes. This Tribbles member induces apoptosis of cells mainly of the hematopoietic origin. It has been identified as a protein up-regulated by inflammatory stimuli in myeloid (THP-1) cells, and also as an oncogene that inactivates the transcription factor C/EBPalpha (CCAAT/enhancer-binding protein alpha) and causes acute myelogenous leukemia. Alternatively spliced transcript variants have been found for this gene.Spécification
TRIB2 | |
Polyclonal | |
Unconjugated | |
TRIB2 | |
C5FW, GS3955, TRB 2, TRB2, TRIB2, Tribbles homolog 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217410, 28951, 313974 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4K3, Q92519 | |
TRIB2 | |
TRIB2 Fusion Protein Ag7605 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |