Learn More
TRIM Antibody (488806), PerCP, Novus Biologicals™
Code produit. 18647953 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantité unitSize
18647953 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit. 18647953 Fournisseur Novus Biologicals Code fournisseur FAB4708C
Mouse Monoclonal Antibody
TRIM Monoclonal antibody specifically detects TRIM in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY
Spécification
Antigène TRIM
Applications Western Blot, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 488806
Conjugué PerCP
Dilution Western Blot, Intracellular Staining by Flow Cytometry
Formule PBS
Alias de gène pp29/30, T cell receptor associated transmembrane adaptor 1, T cell receptor interacting molecule, T-cell receptor interacting molecule, T-cell receptor-associated transmembrane adapter 1, T-cell receptor-interacting molecule, TCRIM, TRIMHSPC062
Espèces hôtes Mouse
Immunogène E. coli-derived recombinant human TRIM, Thr50-Asn186, Accession # Q6PIZ9
Méthode de purification Protein A or G purified from hybridoma culture supernatant
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Adaptive Immunity, Immunology
Primaire ou secondaire Primary
Identification génétique (Entrez) 50852
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG1