missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM32 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10326-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.Spécification
TRIM32 | |
Polyclonal | |
Unconjugated | |
TRIM32 | |
72 kDa Tat interacting protein, BBS11, HT2A, LGMD2H, TATIP, TRIM32, tripartite motif containing 32, Zinc finger protein HT2A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22954, 313264, 69807 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13049, Q8CH72 | |
Trim32 | |
TRIM32 Fusion Protein Ag0401 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |