missing translation for 'onlineSavingsMsg'
Learn More

TRIM38 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13405-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831604

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.
Spécification

Spécification

TRIM38
Polyclonal
Unconjugated
TRIM38
RING finger protein 15, RNF15, RORET, TRIM38, tripartite motif containing 38, Zinc finger protein RoRet
Rabbit
Antigen Affinity Chromatography
RUO
10475
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00635
TRIM38
TRIM38 Fusion Protein Ag3840
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions