Learn More
TRIM59 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28575-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TRIM59 belongs to the tripartite motif (TRIM) family. It functions as E3 ubiquitin ligases or an adaptor protein, and plays important roles in various types of human cancers.
Spécification
| TRIM59 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 286827 | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat |