missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10786-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
TRIM9 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to cytoplasmic bodies. Its function has not been identified. Alternate splicing of this gene generates two transcript variants encoding different isoforms.Spécification
TRIM9 | |
Polyclonal | |
Unconjugated | |
TRIM9 | |
KIAA0282, RING finger protein 91, RNF91, SPRING, TRIM9, tripartite motif containing 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114088, 155812, 94090 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8C7M3, Q91ZY8, Q9C026 | |
TRIM9 | |
TRIM9 Fusion Protein Ag1238 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |