Learn More
TSPAN15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10460-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.Spécification
TSPAN15 | |
Polyclonal | |
Unconjugated | |
TSPAN15 | |
2700063A19Rik, NET 7, NET7, Tetraspan NET 7, tetraspanin 15, TM4SF15, Tspan 15, TSPAN15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23555 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O95858 | |
TSPAN15 | |
TSPAN15 Fusion Protein Ag0710 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |