missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 349.00€
Spécification
Antigène | TSPAN15 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16806863
|
Proteintech
10460-1-AP-20UL |
20 μL |
129.00€
20µL |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16896853
|
Proteintech
10460-1-AP-150UL |
150 μL |
349.00€
150µL |
Expédition estimée: 30-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
TSPAN15 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. The use of alternate polyadenylation sites has been found for this gene.Spécification
TSPAN15 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O95858 | |
23555 | |
TSPAN15 Fusion Protein Ag0710 | |
Primary | |
-20°C | |
TSPAN15 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
2700063A19Rik, NET 7, NET7, Tetraspan NET 7, tetraspanin 15, TM4SF15, Tspan 15, TSPAN15 | |
TSPAN15 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |