missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTYH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26973-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
TTYH1 is a member of the tweety family of proteins, a family of chloride anion channels containing five transmembrane regions. TTYH1 is a Ca2+-independent, volume-sensitive large conductance chloride (Cl-) channel. TTYH1 is primarily expressed in neural tissue and upregulated in astrocytoma, glioma, and several other cancers. Recent experiments have shown that TTYH1 is an integral endoplasmic reticulum (ER) membrane protein involved in cell proliferation and is thought to play an essential role in embryonic cell growth, possibly through the Ca2+ storage/release process in ER membranes during early development.Spécification
TTYH1 | |
Polyclonal | |
Unconjugated | |
TTYH1 | |
hTTY1, Protein tweety homolog 1, TTYH1, tweety homolog 1 (Drosophila) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
292597, 57348, 57776 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P0C5X8, Q9D3A9, Q9H313 | |
Ttyh1 | |
TTYH1 Fusion Protein Ag25290 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |