missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUSC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16039-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certain facial and developmental defects. WRN is a DNA helicase encoded by chromosome 8 and shown defective in those with the early aging disorder Werner syndrome. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.Spécification
TUSC3 | |
Polyclonal | |
Unconjugated | |
TUSC3 | |
M33, MRT7, N33, OST3A, Protein N33, tumor suppressor candidate 3, TUSC3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290783, 7991, 80286 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q13454, Q8BTV1 | |
Tusc3 | |
TUSC3 Fusion Protein Ag8912 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |