missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25687-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
May act as a redox regulator involved in DUOX proteins folding. The interaction with DUOX1 and DUOX2 suggest that it belongs to a multiprotein complex constituting the thyroid H2O2generating system. It is however not sufficient to assist DUOX1 and DUOX2 in H2O2 generation.Spécification
TXNDC11 | |
Polyclonal | |
Unconjugated | |
TXNDC11 | |
EF hand binding protein 1, EFP1, TXNDC11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51061 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PKC3 | |
TXNDC11 | |
TXNDC11 Fusion Protein Ag22618 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |