missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TXNDC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 19834-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein-disulfide isomerase. Its expression is induced by hypoxia and its role may be to protect hypoxic cells from apoptosis. Two transcript variants encoding different isoforms have been found for this gene.Spécification
TXNDC5 | |
Polyclonal | |
Unconjugated | |
TXNDC5 | |
EndoPDI, ER protein 46, ERP46, Hcc 2, PDIA15, Thioredoxin like protein p46, TLP46, TXNDC5, UNQ364 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100362805, 105245, 81567 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8NBS9, Q91W90 | |
Txndc5 | |
TXNDC5 Fusion Protein Ag13808 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |