missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U1A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-58268
This item is not returnable.
View return policy
Description
U1A Polyclonal specifically detects U1A in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| U1A | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| Mud1, small nuclear ribonucleoprotein polypeptide A, U1 snRNP A, U1 snRNP-specific protein A, U1-AU1 small nuclear ribonucleoprotein A, U1AU1 small nuclear RNP-specific A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| SNRPA | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:PGLAPGQIPPGAMPPQQLMPGQMPPAQPLSENPPNHILFLTNLP | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6626 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction