missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBC9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10070-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
In heart and muscle extracts the UBE2I antibodies recognizes a 38 kDa protein band, but this band is not visible in extracts of other rat tissues.
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. This gene encodes a member of the E2 ubiquitin-conjugating enzyme family. Four alternatively spliced transcript variants encoding the same protein have been found for this gene.Spécification
UBC9 | |
Polyclonal | |
Unconjugated | |
UBE2I | |
C358B7.1, E2,UBE2I, P18, SUMO conjugating enzyme UBC9, SUMO protein ligase, UBC9, UBCE9, UBE2I, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, Ubiquitin protein ligase I | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22196, 25573, 7329 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63279, P63280, P63281 | |
Ube2i | |
UBC9 Fusion Protein Ag0111 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |