missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2Q2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12581-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Catalyzes the covalent attachment of ubiquitin to other proteins.Spécification
UBE2Q2 | |
Polyclonal | |
Unconjugated | |
UBE2Q2 | |
UBCi, UBE2Q2, Ubiquitin carrier protein Q2, Ubiquitin protein ligase Q2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109161, 363065, 92912 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K2Z8, Q8WVN8 | |
Ube2q2 | |
UBE2Q2 Fusion Protein Ag3276 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |