missing translation for 'onlineSavingsMsg'
Learn More

Ubiquilin 1 Rabbit anti-Human, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 23516-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16832375

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody detects 70kD band of UBQLN1.

PLIC1 is an ubiquitin-like protein (ubiquilin) that shares high degree of similarity with related products in yeast, rat and frog. Ubiquilins contain a N-terminal ubiquitin-like domain and a C-terminal ubiquitin-associated domain. They physically associate with both proteasomes and ubiquitin ligases, and thus are thought to functionally link the ubiquitination machinery to the proteasome to affect in vivo protein degradation. This ubiquilin has also been shown to modulate accumulation of presenilin proteins, and is found in lesions associated with Alzheimer′s and Parkinson′s disease.
Spécification

Spécification

Ubiquilin 1
Polyclonal
Unconjugated
UBQLN1
DA41, DSK2, hPLIC 1, PLIC 1, PLIC1, ubiquilin 1, UBQLN1, XDRP1
Rabbit
Antigen Affinity Chromatography
RUO
114590, 29979
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JJP9, Q9UMX0
Ubqln1
Ubiquilin 1 Fusion Protein Ag20252
150 μL
Primary
Rat, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions