Learn More
UBR4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 15966-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835384
  • 140.00€ / 20µL
Expédition estimée: 03-05-2024
Description
The UBR4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBR4. This antibody recognizes human, mouse antigen. The UBR4 antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
Spécification
UBR4
Polyclonal
Unconjugated
20 μL
23352
IgG
Immunohistochemistry, ELISA
213 μg/mL
Rabbit
Primary
Human, Mouse