Learn More
UBR4 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15966-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The UBR4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human UBR4. This antibody recognizes human, mouse antigen. The UBR4 antibody has been validated for the following applications: IHC, ELISA analysis.
Spécification
UBR4 | |
Polyclonal | |
Unconjugated | |
20 μL | |
23352 | |
IgG |
Immunohistochemistry, ELISA | |
213 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |